Order Entry
Belgium
ContactUsLinkComponent
Leukotriene A4 ELISA Kit
Leukotriene A4 ELISA Kit
  ANTIA319462-96
New Product
 :  
Leukotriene A4 ELISA Kit
  ANTIA319462-96
New Product
 :  

 

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Other
  • Target protein:
    Leukotriene A4
  • Description:
    Leukotriene A4 ELISA kit
  • Sample type:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Cross reactivity:
    Leukotriene A4 ELISA Kit exhibits high specificity and excellent specificity for the detection of universal Leukotriene A4. No significant cross-reactivity or interference between Leukotriene A4 and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    2 hours
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    15,625 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Sample volume:
    50 μl
  • Sensitivity:
    9,375 ng/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

 

 

Leukotriene A4 ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Leukotriene A4 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay Precision: Intra-Assay: CV <8%, Inter-Assay: CV <10%

Leukotriene A4 ELISA Kit employs the competitive enzyme immunoassay technique for the quantitative measurement of Leukotriene A4 in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with Leukotriene A4 antigen. During the incubation, Leukotriene A4 present in the samples or standards competes with the fixed amount of immobilised Leukotriene A4 for binding sites on the Biotinylated Anti-Leukotriene A4 Antibody. The more Leukotriene A4 present in a sample or standard, the less Biotinylated Anti-Leukotriene A4 Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-Leukotriene A4 Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of Leukotriene A4 present in each sample or standard. The concentration of Leukotriene A4 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a Ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch and protocol manual.
 : This product is for Research use only. It is not for use in diagnostic or therapeutic procedures.