Order Entry
Belgium
ContactUsLinkComponent
 
Mouse G-CSF ELISA Kit (Small Sample Volume)
  ANTIA323208-96
New Product
 :  
Mouse G-CSF ELISA Kit (Small Sample Volume)
  ANTIA323208-96
New Product
 :  

 

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    G-CSF
  • Description:
    Mouse G-CSF ELISA kit (small sample volume)
  • Sample type:
    Serum, plasma, and other biological fluids
  • Cross reactivity:
    No significant cross-reactivity or interference between mouse G-CSF and analogues was observed.
  • Detection method:
    Colorimetric
  • Time to Results:
    3 h 30 min
  • Shelf life:
    Store for 6 months at 4 °C
  • Detection range:
    15,62 - 1000 pg/ml
  • Storage temperature:
    4 °C
  • Sensitivity:
    8,38 pg/ml
  • Regulatory status:
    RUO
  • Tests per kit:
    96 tests

 

 

Mouse G-CSF ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse G-CSF in serum, plasma, and other biological fluids. This ELISA kit only requires 25 µl of sample volume(s).

  • Small Sample Volume: Requires as little as 25 µl of sample!
  • Assay Precision: Intra-Assay: CV <10%, Inter-Assay: CV <10%

Mouse G-CSF ELISA kit employs the sandwich Enzyme-Linked Immunoassay technique for the quantitative measurement of mouse G-CSF in serum, plasma, and other biological fluids. An antibody specific for G-CSF has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the G-CSF present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-G-CSF Antibody, which binds the captured G-CSF present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of G-CSF captured in each well. The concentration of G-CSF can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.