Orders LinkContactUsLinkComponent
Filtration funnels, cylindrical
Filtration funnels, cylindrical
Supplier:   SciLabware
Filtration funnels, cylindrical
Supplier:  SciLabware
Certificates
Filtration funnels, cylindrical
Filtration funnels, cylindrical
5 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass, these are ideal for filtering precipitates.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With integrated sintered glass filter plate.
Product Family Options
Product Information
  • Capacity
    Length
    Plate Ø
    Porosity
    Availability
    Price
  • 50 ml
    160 mm
    20 mm
    1
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 563,00 €
    563,00 €
    1 * 10 ST
       563,00 €
  • 50 ml
    160 mm
    20 mm
    2
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 563,00 €
    563,00 €
    1 * 10 ST
       563,00 €
  • 50 ml
    160 mm
    20 mm
    3
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 563,00 €
    563,00 €
    1 * 10 ST
       563,00 €
  • 50 ml
    160 mm
    20 mm
    4
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 563,00 €
    563,00 €
    1 * 10 ST
       563,00 €
  • 50 ml
    160 mm
    20 mm
    5
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 614,00 €
    614,00 €
    1 * 10 ST
       614,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.