Orders LinkContactUsLinkComponent
Faeces containers
Faeces containers
Marketsource
Supplier:   SARSTEDT
Faeces containers
Supplier:  SARSTEDT
Marketsource
Certificates
Faeces containers
11 Options Available Below
Product Details & Documents
Containers with sampling scoops enables clean and easy collection of faeces samples.
  • To collect defined faeces volume of 1 ml, approximately 1 g
  • Sterile or non sterile
  • Labelled or opaque options available
Product Family Options
Product Information
  • Type
    Material
    Ext.Ø
    Height
    Availability
    Price
  • Sterile, with flat base and push cap
    PS
    23,5 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 148,39 €
    148,39 €
    1 * 250 ST
       148,39 €
    Specifications:
    Marketsource
    Marketsource
  • Non-sterile, with flat base and push cap
    PS
    23,5 mm
    75 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 161,59 €
    161,59 €
    1 * 500 ST
       161,59 €
    Specifications:
    Marketsource
    Marketsource
  • Sterile, with screw cap
    PP
    20 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 99,88 €
    99,88 €
    1 * 500 ST
       99,88 €
    Specifications:
    Marketsource
    Marketsource
  • Non-sterile, with screw cap
    PP
    20 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 89,32 €
    89,32 €
    1 * 500 ST
       89,32 €
    Specifications:
    Marketsource
    Marketsource
  • Non-sterile, with screw cap, with label
    PP
    20 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 104,29 €
    104,29 €
    1 * 500 ST
       104,29 €
    Specifications:
    Marketsource
    Marketsource
  • Sterile, with screw cap, with label
    PP
    20 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 167,78 €
    167,78 €
    1 * 500 ST
       167,78 €
    Specifications:
    Marketsource
    Marketsource
  • Non-sterile, with screw cap, with label
    PP, white
    20 mm
    76 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 159,46 €
    159,46 €
    1 * 500 ST
       159,46 €
    Specifications:
    Marketsource
    Marketsource
  • Sterile, with screw cap
    PP
    16,5 mm
    101 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 98,96 €
    98,96 €
    1 * 500 ST
       98,96 €
    Specifications:
    Marketsource
    Marketsource
  • Non-sterile, with screw cap
    PP
    16,5 mm
    101 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 85,99 €
    85,99 €
    1 * 500 ST
       85,99 €
    Specifications:
    Marketsource
    Marketsource
  • Sterile, with screw cap
    PP
    25 mm
    107 mm
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 146,48 €
    146,48 €
    1 * 400 ST
       146,48 €
    Specifications:
    Marketsource
    Marketsource
  • Non-sterile, with screw cap
    PP
    25 mm
    107 mm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 120,68 €
    120,68 €
    1 * 500 ST
       120,68 €
    Specifications:
    Marketsource
    Marketsource