Orders LinkContactUsLinkComponent
Container with screw cap
Marketsource
Supplier:   SARSTEDT
Container with screw cap
Supplier:  SARSTEDT
Marketsource
Certificates
Container with screw cap
10 Options Available Below
Product Details & Documents
PP, translucent, with screw cap.
  • Excellent chemical resistance
  • Excellent temperature resistance
Transparent containers with reliable sealing caps.
Collection and storage of urine.
 : *The product(s) on this page are outside the scope of the EU Directive 2019/904
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Type
    Cap colour
    Packed
    Material
    Availability
    Price
  • 70 ml
    44 mm
    55 mm
    HDPE cap, with label, sterile, individually wrapped
    Yellow
    100/pack
    PP
    Log in for availability
    Log in for availability
    1 * 200 ST
    1 * 200 ST - 104,29 €
    104,29 €
    1 * 200 ST
       104,29 €
    Specifications:
    Marketsource
    Marketsource
  • 70 ml
    44 mm
    55 mm
    HDPE cap
    Yellow
    500/case
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 97,72 €
    97,72 €
    1 * 500 ST
       97,72 €
    Specifications:
    Marketsource
    Marketsource
  • 70 ml
    44 mm
    55 mm
    HDPE cap with label
    Yellow
    500/case
    PP
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 110,39 €
    110,39 €
    1 * 500 ST
       110,39 €
    Specifications:
    Marketsource
    Marketsource
  • 120 ml
    44 mm
    105 mm
    PP cap, without graduation
    Neutral
    250/case
    PC
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST
    Specifications:
    Marketsource
    Marketsource
  • 120 ml
    44 mm
    105 mm
    HDPE cap, sterile, graduation upto 120 ml
    Yellow
    250/case
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 98,25 €
    98,25 €
    1 * 250 ST
       98,25 €
    Specifications:
    Marketsource
    Marketsource
  • 120 ml
    44 mm
    105 mm
    HDPE cap, graduation upto 120 ml
    Neutral
    250/case
    PP
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 96,29 €
    96,29 €
    1 * 250 ST
       96,29 €
    Specifications:
    Marketsource
    Marketsource
  • 250 ml
    70 mm
    78 mm
    PP cap, graduation upto 200 ml
    Neutral
    210/case
    PC
    Log in for availability
    Log in for availability
    1 * 210 ST
    1 * 210 ST
    Specifications:
    Marketsource
    Marketsource
  • 250 ml
    70 mm
    78 mm
    HDPE cap, graduation upto 200 ml
    Neutral
    210/case
    PP
    Log in for availability
    Log in for availability
    1 * 75 ST
    1 * 75 ST - 51,38 €
    51,38 €
    1 * 75 ST
       51,38 €
  • 250 ml
    70 mm
    78 mm
    HDPE cap, sterile, graduation up to 200 ml
    Yellow
    210/case
    PP
    Log in for availability
    Log in for availability
    1 * 150 ST
    1 * 150 ST - 120,04 €
    120,04 €
    1 * 150 ST
       120,04 €
    Specifications:
    Marketsource
    Marketsource
  • 500 ml
    70 mm
    150 mm
    HDPE cap, sterile, graduation up to 420 ml
    Yellow
    105/case
    PP
    Log in for availability
    Log in for availability
    1 * 70 ST
    1 * 70 ST - 121,33 €
    121,33 €
    1 * 70 ST
       121,33 €
    Specifications:
    Marketsource
    Marketsource