ContactUsLinkComponent
Recombinant protein purification resins, IMAC
Recombinant protein purification resins, IMAC
Supplier:  G-Biosciences
Certificates
undefined
Recombinant protein purification resins, IMAC
Supplier:  G-Biosciences
Specifications
  • Binding capacity:
    >50 mg/ml
  • Ligand density:
    20 - 40 μmoles Cu2+/ml
  • Description:
    Cobalt chelating resin, 1 ml columns, for the purification of 6X His proteins