Orders LinkContactUsLinkComponent
Solid phase extraction cartridges, Bond Elut™ Plexa PCX
Solid phase extraction cartridges, Bond Elut™ Plexa PCX
Supplier:   VARIAN
Solid phase extraction cartridges, Bond Elut™ Plexa PCX
Supplier:  VARIAN
Certificates
Solid phase extraction cartridges, Bond Elut™ Plexa PCX
11 Options Available Below
Product Details & Documents
Bond Elut™ Plexa PCX uses a polymeric cation exchange resin that combines the outstanding properties of Bond Elut™ Plexa - superior flow characteristics and improved analytical performance - with strong cation exchange functionalities. This mixed-mode SPE sorbent removes neutral and acidic interferences from the matrix, concentrates basic analytes and therefore improves sensitivity in the determination of basic compounds.
  • Faster flow rates improve productivity
  • Extraction cleanliness and reduced interference improve precision
  • Simplified, single method
Specifications for Product Family
Specifications
  • Catalog No:
  • VARI12108301B
  • VARI12108301
  • 549-0808
  • VARI12108303B
  • VARI12108303
  • 549-0809
  • 549-0810
  • VARI12108206
  • 549-0838
  • VARI12281306
  • VARI5982-0603
  • Surface area:
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
  • 550 m²/g
Specifications
Product Family Options
Straight barrel cartridges
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12108301B
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    1 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 1.000 ST
    1 * 1.000 ST - 2 820,00 €
    2 820,00 €
    1 * 1.000 ST
       2 820,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12108301B
    Surface area
    550 m²/g
  • VARI12108301
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    1 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 299,00 €
    299,00 €
    1 * 100 ST
       299,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12108301
    Surface area
    550 m²/g
  • 549-0808
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    1 ml
    60 mg
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 304,00 €
    304,00 €
    1 * 100 ST
       304,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0808
    Surface area
    550 m²/g
  • VARI12108303B
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 370,00 €
    1 370,00 €
    1 * 500 ST
       1 370,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12108303B
    Surface area
    550 m²/g
  • VARI12108303
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    3 ml
    30 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 155,00 €
    155,00 €
    1 * 50 ST
       155,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12108303
    Surface area
    550 m²/g
  • 549-0809
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    3 ml
    60 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 185,00 €
    185,00 €
    1 * 50 ST
       185,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0809
    Surface area
    550 m²/g
  • 549-0810
    Straight barrel cartridge, tabless
    Straight barrel cartridge, tabless
    Plexa PCX
    100 Å
    45 µm
    3 ml
    60 mg
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 183,00 €
    183,00 €
    1 * 50 ST
       183,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0810
    Surface area
    550 m²/g
  • VARI12108206
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    6 ml
    200 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 247,00 €
    247,00 €
    1 * 30 ST
       247,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12108206
    Surface area
    550 m²/g
  • 549-0838
    Straight barrel cartridge
    Straight barrel cartridge
    Plexa PCX
    100 Å
    45 µm
    6 ml
    500 mg
    Log in for availability
    Log in for availability
    1 * 30 ST
    1 * 30 ST - 367,00 €
    367,00 €
    1 * 30 ST
       367,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    549-0838
    Surface area
    550 m²/g
Other formats
Product Information
  • Phase
    Pore size
    Particle Size
    Capacity
    Bed weight
    Availability
    Price
  • VARI12281306
    800 series cartridge
    800 series cartridge
    Plexa PCX
    100 Å
    45 µm
    Log in for availability
    Log in for availability
    1 * 96 ST
    1 * 96 ST - 1 080,00 €
    1 080,00 €
    1 * 96 ST
       1 080,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI12281306
    Surface area
    550 m²/g
  • VARI5982-0603
    LRC cartridge
    LRC cartridge
    Plexa PCX
    100 Å
    45 µm
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 1 470,00 €
    1 470,00 €
    1 * 500 ST
       1 470,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    VARI5982-0603
    Surface area
    550 m²/g
Bond Elut™ 96 round well plates
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 30 mg
    96-round well plate for SPE, Plexa™ PCX
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 436,00 €
    436,00 €
    1 * 1 ST
       436,00 €
Product Information
  • Catalog #
    Filling
    Description
    Availability
    Price
  • 10 mg
    96-round well plate for SPE, Plexa™ PCX
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 446,00 €
    446,00 €
    1 * 1 ST
       446,00 €
Bond Elut™ 96 square well plates
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • 96-square well plate for SPE, 2 ml, Plexa™ PCX
    10 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 446,00 €
    446,00 €
    1 * 1 ST
       446,00 €
Product Information
  • Catalog #
    Description
    Filling
    Availability
    Price
  • 96-square well plate for SPE, 2 ml, Plexa™ PCX
    30 mg
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 464,00 €
    464,00 €
    1 * 1 ST
       464,00 €