ContactUsLinkComponent
Aspirating pipettes
Catalog # SARS86.1252.011
Supplier:  SARSTEDT
Aspirating pipettes
Catalog # SARS86.1252.011
Supplier:  SARSTEDT
Specifications
  • Capacity:
    2 ml
  • Environmentally Preferable:
  • Pk:
    1.000
Specifications
About this item
PS pipettes, non graduated, unplugged, sterile.
  • For aspirating media with a vacuum pump
  • Certified non pyrogenic
  • Individually wrapped in paper plastic peel packaging