Membrane filters, Millipore Express® PLUS
Supplier: Merck
Certificates
About this item
These PES white membrane filters provides ultrafast filtration of tissue culture media, additives, buffers and other aqueous solutions. This high throughput, low protein-binding membrane is also used in many of our ready-to-use sterile filtration devices.
- Ultra-fast filtration and high throughput shorten process time
- Can be sterilised by autoclave, ethylene oxide, or gamma irradiation
Widely used for sterile filtration, buffer filtration and tissue culture media filtration.
Surface: plain
Bacterial endotoxins: 0,5 EU/ml
Gravimetric extractables: <0,5%
Specifications
- Catalog No:
- MILFGPWP02500
- MILFGPWP01300
- MILFHPWP09050
- MILFGPWP04700
- MILFGPWP09050
- MILFHPWP14250
- MILFHPWP01300
- MILFGPWP14250
- MILFHPWP02500
- MILFHPWP04700
- Type:
- GPWP
- GPWP
- HPWP
- GPWP
- GPWP
- HPWP
- HPWP
- GPWP
- HPWP
- HPWP
- Pore size:
- 0,22 µm
- 0,22 µm
- 0,45 µm
- 0,22 µm
- 0,22 µm
- 0,45 µm
- 0,45 µm
- 0,22 µm
- 0,45 µm
- 0,45 µm
- Thickness:
- 170 µm
- 170 µm
- 140 µm
- 170 µm
- 170 µm
- 140 µm
- 140 µm
- 170 µm
- 140 µm
- 140 µm
- Water flow rate:
- 15 ml/min/cm² (690 mbar)
- 15 ml/min/cm² (690 mbar)
- 23 ml/min/cm² (690 mbar)
- 15 ml/min/cm² (690 mbar)
- 15 ml/min/cm² (690 mbar)
- 23 ml/min/cm² (690 mbar)
- 23 ml/min/cm² (690 mbar)
- 15 ml/min/cm² (690 mbar)
- 23 ml/min/cm² (690 mbar)
- 23 ml/min/cm² (690 mbar)
- Airflow rate:
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
- 40 ml/min cm²
Specifications
Frequently Bought Together