ContactUsLinkComponent
Centrifuge tubes, pear shape, ASTM, PYREX®
  BIBB3440/02M
undefined
Centrifuge tubes, pear shape, ASTM, PYREX®
  BIBB3440/02M
  • Closure type:
    Without cap
  • Ø×L:
    17×157 mm
  • Capacity:
    100 ml
  • Environmentally Preferable:
  • Pk:
    2
Centrifuge tube is used for the determination of water and sediment in petroleum products.
  • Graduated in white enamel
Lower stem graduated from 0 to 1,5 ml in 0,1 ml divisions whereas upper body graduated from 1,5 to 5 ml in 0,5 ml divisions and from 5 to 10 ml in 1 ml.
 : Complies with ASTM D96 and D-1966.