Orders LinkContactUsLinkComponent
Cavity slides
Cavity slides
Cavity slides
Certificates
Cavity slides
4 Options Available Below
Product Details & Documents
Cavity microscopy slides.
  • Ground edges
  • Made from clear glass
Suitable for staining applications in microscopy.
Product Family Options
Product Information
  • Colour
    No. of wells
    Thickness
    Well Ø
    Availability
    Price
  • HECH42410010
    Cavity slides
    Cavity slides
    Clear
    1
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 129,00 €
    129,00 €
    1 * 50 ST
       129,00 €
  • 631-9489
    Cavity slides
    Cavity slides
    Clear
    2
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 179,00 €
    179,00 €
    1 * 50 ST
       179,00 €
  • HECH42412010
    Cavity slides
    Cavity slides
    Clear
    3
    1,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 260,00 €
    260,00 €
    1 * 50 ST
       260,00 €
  • HECH42415010
    Cavity slides
    Cavity slides
    Clear
    1
    3 - 4 mm
    22 mm
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 207,00 €
    207,00 €
    1 * 10 ST
       207,00 €