Microcentrifuge tubes, SafeSeal
: SARSTEDT
PP, translucent.
- Wide hinge for easy and precise closure
- Large writing space on lid, graduated
- Centrifugation to 30000×g (2,0 ml tubes: to 25000×g)
For transport, storage and sample processing in the laboratory. DNA-/RNase-free tubes and screw caps for the storage of high-grade synthesised oligonucleotides, enzymes, buffers etc. Retaining cams ensure a safe fit also under thermal strain.
Frequently Bought Together