ContactUsLinkComponent
Adapters, pig receiver, Quickfit®
Adapters, pig receiver, Quickfit®
Catalog # 271-0013
Supplier:  SciLabware
undefined
Adapters, pig receiver, Quickfit®
Catalog # 271-0013
Supplier:  SciLabware
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    14/23 NS
  • Pk:
    1
Specifications
About this item
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Allows up to three flasks to be connected to a condenser outlet. Useful for collecting distillate fractions. By slightly rotating the adapter a new flask is presented to the distillate. With side-arm for connection to a vacuum supply.