Orders LinkContactUsLinkComponent
Adapters, receiver, with side socket, Quickfit®
Adapters, receiver, with side socket, Quickfit®
14/23 NS, 14/23 NS
Catalog # 271-0012
Supplier:   SciLabware
Adapters, receiver, with side socket, Quickfit®
Catalog # 271-0012
Supplier:  SciLabware
Adapters, receiver, with side socket, Quickfit®
Adapters, receiver, with side socket, Quickfit®
14/23 NS, 14/23 NS
Price
69,50 €
Log in for availability
1 * 1 ST
Product Details & Documents
PYREX®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With side socket inclined at an angle of 105°.
Specifications
  • Cone size:
    14/23 NS
  • Socket size:
    14/23 NS