Adapters with stopcock and socket
Supplier: SciLabware
Certificates
About this item
PYREX®, borosilicate glass, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
With GP Rotaflo® PTFE stopcock providing fine flow control and positive closure. Detachable plastic screwthread connectors allow rubber hose etc. to be fitted easily and safely .
Frequently Bought Together