ContactUsLinkComponent
Closures for square bottles, with tamper-evident ring
Closures for square bottles, with tamper-evident ring
Catalog # KJSKRK42
undefined
Closures for square bottles, with tamper-evident ring
Catalog # KJSKRK42
Specifications
  • Thread:
    50 mm
  • Pk:
    1
  • For bottles:
    250/500/750/1000 ml
Specifications
About this item
HDPE, black.
  • Good chemical resistance
  • Good temperature resistance