Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Pipette tips

Marketsource
Supplier:   SARSTEDT
Pipette tips
Supplier:  SARSTEDT
Marketsource
Certificates

22 Options Available Below

Product Details & Documents

These high quality tips are suitable for the most common pipettes.

  • Sterile tips are DNase/RNase- and ATP-free, and non pyrogenic
  • Available loose in bags, packed in racks or in Tip StackPack reload systems

Tip StackPack: Tips are preloaded in reusable, stacked 96-tip trays. The trays are designed to safely cover each individual 96-tip layer, shielding the preloaded tips in the stack like a protective screen.

Product Family Options

Product Information

  • Tip style
    Volume
    Sterile
    Tip colour
    For use with
    Packed
    Availability
    Price
  • Type A
    200 µl
    +
    Yellow
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical
    96/box, 480/inner case, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type B
    1000 µl
    -
    Blue
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    100/box, 1000/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST - 84,69 €
    84,69 €
    1 * 1.920 ST
       84,69 €

    Specifications:
    Marketsource
    Marketsource
  • Type B
    1000 µl
    +
    Blue
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    100/box, 500/inner case, 1000/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST - 137,76 €
    137,76 €
    1 * 1.920 ST
       137,76 €

    Specifications:
    Marketsource
    Marketsource
  • Type B
    1000 µl
    -
    Blue
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    250/bag, 5000/case
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST - 100,90 €
    100,90 €
    1 * 5.000 ST
       100,90 €

    Specifications:
    Marketsource
    Marketsource
  • Type C
    250 µl
    -
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical
    500/bag, 10000/case
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST

    Specifications:
    Marketsource
    Marketsource
  • Type C
    250 µl
    +
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical
    96/box, 480/inner case, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type C
    250 µl
    -
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical
    Tip stack pack, 96/tray, 480/stack pack, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type D
    10 µl
    -
    Transparent
    Eppendorf Reference, Research and Research Plus
    1000/bag, 10000/case
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST

    Specifications:
    Marketsource
    Marketsource
  • Type E
    10 µl
    -
    Transparent
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    96/box, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST - 84,49 €
    84,49 €
    1 * 1.920 ST
       84,49 €

    Specifications:
    Marketsource
    Marketsource
  • Type E
    10 µl
    +
    Transparent
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    96/box, 480/inner case, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type E
    10 µl
    -
    Transparent
    Gilson, Eppendorf, Finnpipette, Biohit, Brand and Socorex
    Tip stack pack, 96/tray, 576/stack pack, 2304/case
    Log in for availability
    Log in for availability
    1 * 2.304 ST
    1 * 2.304 ST

    Specifications:
    Marketsource
    Marketsource
  • Type E
    10 µl
    -
    Transparent
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    Tip stack pack, 96/tray, 576/stack pack, 2304/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST - 70,49 €
    70,49 €
    1 * 1.920 ST
       70,49 €

    Specifications:
    Marketsource
    Marketsource
  • Type E
    10 µl
    -
    Transparent
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical, eLine Electronic
    1000/bag, 10000/case
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST

    Specifications:
    Marketsource
    Marketsource
  • Type E, long
    10 µl
    -
    Transparent
    Gilson
    96/box, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type E, long
    10 µl
    -
    Transparent
    Gilson
    500/bag, 10000/case
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST

    Specifications:
    Marketsource
    Marketsource
  • Type J
    20 µl
    +
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
    96/box, 480/inner case, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type J
    20 µl
    -
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
    1000/bag, 10000/case
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST

    Specifications:
    Marketsource
    Marketsource
  • Type L
    300 µl
    -
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
    96/box, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type L
    300 µl
    +
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
    96/box, 480/inner case, 1920/case
    Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST

    Specifications:
    Marketsource
    Marketsource
  • Type L
    300 µl
    -
    Transparent
    Eppendorf Reference, Research/Research Plus, mLine Mechanical, Proline Mechanical, eLine Electronic
    500/bag, 5000/case
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST

    Specifications:
    Marketsource
    Marketsource
  • Type M
    1250 µl
    -
    Transparent
    Eppendorf Reference, Research, mLine Mechanical, Proline Mechanical
    96/box, 3840/case
    Log in for availability
    Log in for availability
    1 * 3.840 ST
    1 * 3.840 ST

    Specifications:
    Marketsource
    Marketsource
  • Log in for availability
    Log in for availability
    1 * 1.920 ST
    1 * 1.920 ST - 67,42 €
    67,42 €
    1 * 1.920 ST
       67,42 €

    Specifications:
    Marketsource
    Marketsource
Accessories

Product Information

  • Catalog #
    Colour
    Availability
    Price
  • Tip system box for pipette tip trays, with hinged lid, empty
    Tip system box for pipette tip trays, with hinged lid, empty
    SARS95.1298.001
    Transparent
    Log in for availability
    1 * 1 ST