Orders LinkContactUsLinkComponent
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
Supplier:   SciLabware
Spherical ground joints, Quickfit®
Supplier:  SciLabware
Certificates
Spherical ground joints, Quickfit®
Spherical ground joints, Quickfit®
8 Options Available Below
Product Details & Documents
Made of PYREX®, borosilicate glass.
  • Excellent sealing properties
  • Minimal requirement for joint lubrication
  • Ideal for use as flexible connectors
For the Glassblower who demands the highest quality joint.
Length: 100 mm
Product Family Options
Male, ball joint
Product Information
  • Ball
    Socket
    Availability
    Price
  • S 13/5
    -
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 203,00 €
    203,00 €
    1 * 10 ST
       203,00 €
  • S 19
    -
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 197,00 €
    197,00 €
    1 * 10 ST
       197,00 €
  • S 29
    -
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 269,00 €
    269,00 €
    1 * 10 ST
       269,00 €
  • S 35
    -
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 168,00 €
    168,00 €
    1 * 5 ST
       168,00 €
Female, cup joint
Product Information
  • Ball
    Socket
    Availability
    Price
  • -
    S 13/5
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 187,00 €
    187,00 €
    1 * 10 ST
       187,00 €
  • -
    S 19
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 197,00 €
    197,00 €
    1 * 10 ST
       197,00 €
  • -
    S 29
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 244,00 €
    244,00 €
    1 * 10 ST
       244,00 €
  • -
    S 35
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,00 €
    30,00 €
    1 * 1 ST
       30,00 €