Orders LinkContactUsLinkComponent
Adapters, socket /screwthread
Adapters, socket /screwthread
29/32 NS, SVL 30
Catalog # BIBB670-05
Supplier:   SciLabware
Adapters, socket /screwthread
Catalog # BIBB670-05
Supplier:  SciLabware
Adapters, socket /screwthread
Adapters, socket /screwthread
29/32 NS, SVL 30
Price
768,00 €
Log in for availability
1 * 10 ST
Product Details & Documents
Adapters, borosilicate glass 3.3. Facilitates connection between SVL® and Quickfit® glassware. Features an SVL® screwthread and Quickfit® ground glass socket. Thread accepts SVL® screwcaps and couplings. Socket fits Quickfit® cones.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Socket size:
    29/32 NS
  • Thread:
    SVL 30