Orders LinkContactUsLinkComponent
Accessories for screw thread joints
Accessories for screw thread joints
Supplier:   SciLabware
Accessories for screw thread joints
Supplier:  SciLabware
Certificates
Accessories for screw thread joints
Accessories for screw thread joints
16 Options Available Below
Product Details & Documents
For use with Quickfit® glass screw thread.
Product Family Options
Screw caps
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 6,0 -7,0 mm
    13
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 45,40 €
    45,40 €
    1 * 10 ST
       45,40 €
  • 7,0 - 8,5 mm
    18
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 67,50 €
    67,50 €
    1 * 10 ST
       67,50 €
  • 6,0 - 7,0 mm
    24
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 59,80 €
    59,80 €
    1 * 10 ST
       59,80 €
  • 10,5 - 11,5 mm
    28
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 86,80 €
    86,80 €
    1 * 10 ST
       86,80 €
Silicone rubber ring
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 6,0 - 7,0 mm
    13
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 31,60 €
    31,60 €
    1 * 10 ST
       31,60 €
  • 7,0 - 8,5 mm
    18
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 39,30 €
    39,30 €
    1 * 10 ST
       39,30 €
  • 6,0 - 7,0 mm
    24
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 60,10 €
    60,10 €
    1 * 10 ST
       60,10 €
  • 10,5 - 11,5 mm
    24
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 45,40 €
    45,40 €
    1 * 10 ST
       45,40 €
  • 10,5 - 11,5 mm
    28
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 108,00 €
    108,00 €
    1 * 10 ST
       108,00 €
  • 6,0 - 7,0 mm
    28
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 97,80 €
    97,80 €
    1 * 10 ST
       97,80 €
PTFE washer
Product Information
  • Hole Ø
    Thread
    Availability
    Price
  • 6,0 - 7,0 mm
    13
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 15,00 €
    15,00 €
    1 * 10 ST
       15,00 €
  • 7,0 - 8,5 mm
    18
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 28,60 €
    28,60 €
    1 * 10 ST
       28,60 €
  • 6,0 -7,0 mm
    24
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 44,30 €
    44,30 €
    1 * 10 ST
       44,30 €
  • 10,5 - 11,5 mm
    24
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 32,30 €
    32,30 €
    1 * 10 ST
       32,30 €
  • 10,5 - 11,5 mm
    28
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 39,30 €
    39,30 €
    1 * 10 ST
       39,30 €
  • 18,0 - 19,0 mm
    28
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 40,00 €
    40,00 €
    1 * 10 ST
       40,00 €