Order Entry
Belgium
ContactUsLinkComponent
Distillate receiver Crow, PYREX®
Distillate receiver Crow, PYREX®
Catalog # BIBB3400/04
Supplier:  SciLabware
undefined
Distillate receiver Crow, PYREX®
Catalog # BIBB3400/04
Supplier:  SciLabware

Specifications

  • Capacity:
    100 ml
  • Height:
    250 mm
  • Division:
    1,0 ml
  • Pk:
    2

Specifications

About this item

PYREX®, borosilicate glass, Class B.

  • For the collection of distillate in oil and tar tests
  • Graduations in white enamel
  • Complies with BS 658