Order Entry
Belgium
ContactUsLinkComponent
2-Hydroxy-6-methylnicotinic acid ≥98.0% (by titrimetric analysis)
2-Hydroxy-6-methylnicotinic acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
2-Hydroxy-6-methylnicotinic acid ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light orange to Yellow to Green powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in hot Water:
    very faint turbidity

Specification Test Results

Related Information

Frequently Bought Together