

Palladium(II) nitrate 1% in nitric acid 10%, Specpure® matrix modifier solution
Catalog # 44241.AE
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 mL
Specifications
About this item
:
:230.43 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:3264
ADR:8,III
:GHS05
:GHS07
CAS Number:10102-05-3
Formula:N₂O₆Pd
MDL Number:MFCD00011169