Order Entry
Belgium
ContactUsLinkComponent
Palladium(II) nitrate 0.1% in nitric acid 10%, Specpure® matrix modifier solution
Oxidizing
Corrosive
Palladium(II) nitrate 0.1% in nitric acid 10%, Specpure® matrix modifier solution
Catalog # 44765.AE
CAS Number:  
Oxidizing
Corrosive
Palladium(II) nitrate 0.1% in nitric acid 10%, Specpure® matrix modifier solution
Catalog # 44765.AE
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL

Specifications

About this item

 :
 :230.43 g/mol
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
 :GHS03
 :GHS05
CAS Number:10102-05-3
Formula:N₂O₆Pd
MDL Number:MFCD00011169