VWR®, Niobium standard solution, 1000 mg/L Nb in 5% nitric acid with hydrofluoric acid (max. 1%) AVS TITRINORM standard for AAS
Supplier: VWR Chemicals
Certificates
About this item
- Traceable to SRM from NIST
- Manufactured and tested in ISO Guide 34/ ISO17025 accredited facilities
- Supplied with certificate of analysis, real value ±0,3%
Limited Time Sale
Select items below are currently on sale
Frequently Bought Together