Order Entry
Belgium
ContactUsLinkComponent
 
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAT0974-25G
Supplier:  TCI
CAS Number:  
undefined
TAPS (3-(tris(hydroxymethyl)methylamino)propane-1-sulphonic acid) ≥98.0% (by titrimetric analysis) Good's buffer component for biological research
Catalog # TCIAT0974-25G
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    25 g
  • Seq:
    3308

Specifications

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (Neutralization titration):
    min. 98.0 %
  • Solubility in Water:
    almost transparency

Specification Test Results

About this item

 :
 :243.28 g/mol
Storage Temperature:Ambient
EINECS:249-954-1
CAS Number:29915-38-6
Formula:C₇H₁₇NO₆S
MDL Number:MFCD00007538