Order Entry
Belgium
ContactUsLinkComponent
 
 
 
Bis(triphenylphosphine)nickel(II) chloride ≥96.0% (by titrimetric analysis)
 :  TCI
 
undefined
 
 
Bis(triphenylphosphine)nickel(II) chloride ≥96.0% (by titrimetric analysis)
 :  TCI

 

  • Appearance:
    Dark green to Dark blue to Black powder to crystal
  • Purity (Chelometric Titration):
    min. 96.0 %
  • Purity (Iodometric Titration):
    min. 96.0 %

 

Frequently Bought Together