Order Entry
Belgium
ContactUsLinkComponent
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98.0% (by titrimetric analysis)
Health
Irritant
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98.0% (by titrimetric analysis)
Supplier:  TCI
Certificates
undefined
Health
Irritant
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) ≥98.0% (by titrimetric analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 97.0 area%
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Solubility in hot Water:
    almost transparency
  • Drying loss:
    max. 1.0 %

Specification Test Results

Related Information

Frequently Bought Together