Order Entry
Belgium
ContactUsLinkComponent
Sodium dodecyl sulphate (SDS) ≥97.0% (by GC, titration analysis)
Irritant
Health
Sodium dodecyl sulphate (SDS) ≥97.0% (by GC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
Health
Sodium dodecyl sulphate (SDS) ≥97.0% (by GC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Orange to Green powder to crystaline
  • Purity (GC):
    min. 97.0 %
  • Purity (Neutralization back titration):
    min. 97.0 %
  • Average molecular weight:
    287 to 292

Specification Test Results

Frequently Bought Together