
Glycerine ≥99.0% (by GC) matrix for FABMS and liquid SIMS
Supplier: TCI
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:14,04484
Boiling Pt:290°C
Melting Pt:18.6 °C
Density:1,26 g/cm³ (20 °C)
Flash Pt:177°C
EINECS:200-289-5
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 99.0 %
Specification Test Results
Frequently Bought Together