


Methanol ≥99.8% (by GC) for HPLC
Catalog # TCIAM0628-500ML
Supplier: TCI
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
Specifications
Specification Test Results
- Appearance:Colorless clear liquid
- Purity (GC):min. 99.8 %
- Absorbance (210nm):max. 0.700
- Absorbance (220nm):max. 0.400
- Absorbance (230nm):max. 0.200
Specification Test Results
About this item
:
:32.04 g/mol
Merck Index:14,05957
Boiling Pt:64°C
Melting Pt:–98 °C
Density:0,7918 g/cm³ (20 °C)
Flash Pt:12°C (closed cup)
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
ADR:3,II
REACH:01-2119433307-44
:GHS02
:GHS06
:GHS08
CAS Number:67-56-1
Formula:CH₃OH
MDL Number:MFCD00004595