Order Entry
Belgium
ContactUsLinkComponent
2-Propanol ≥99.5% (by GC) for HPLC
Flammable
Irritant
2-Propanol ≥99.5% (by GC) for HPLC
Catalog # TCIAI0277-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Irritant
2-Propanol ≥99.5% (by GC) for HPLC
Catalog # TCIAI0277-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.5 %
  • Density (20deg.C):
    0.7840 to 0.7870 g/mL
  • Absorbance (210nm):
    max. 0.800
  • Absorbance (220nm):
    max. 0.400

Specification Test Results

About this item

 :
 :60.1 g/mol
Merck Index:14,05208
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0,786 g/cm³ (20 °C)
Flash Pt:15°C
Storage Temperature:Ambient
EINECS:200-661-7
UN:1219
ADR:3,II
REACH:01-2119457558-25
 :GHS02
 :GHS07
CAS Number:67-63-0
Formula:(CH₃)₂CHOH
MDL Number:MFCD00011674