Order Entry
Belgium
ContactUsLinkComponent
1-Butanol ≥99.0% (by GC) for HPLC
Flammable
Corrosive
Irritant
1-Butanol ≥99.0% (by GC) for HPLC
Catalog # TCIAB0944-500ML
Supplier:  TCI
CAS Number:  
undefined
Flammable
Corrosive
Irritant
1-Butanol ≥99.0% (by GC) for HPLC
Catalog # TCIAB0944-500ML
Supplier:  TCI
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

Specifications

Specification Test Results

  • Appearance:
    Colorless clear liquid
  • Purity (GC):
    min. 99.0 %
  • Absorbance (210nm):
    max. 1.000
  • Absorbance (220nm):
    max. 0.500
  • Absorbance (230nm):
    max. 0.250

Specification Test Results

About this item

 :
 :74.12 g/mol
Merck Index:14,01540
Boiling Pt:118°C
Melting Pt:–89 °C
Density:0,812 g/cm³ (20 °C)
Flash Pt:37°C
Storage Temperature:Ambient
EINECS:200-751-6
UN:1120
ADR:3,III
REACH:01-2119484630-38
 :GHS02
 :GHS05
 :GHS07
CAS Number:71-36-3
Formula:H₃C(CH₂)₂CH₂OH
MDL Number:MFCD00002964