

Hydrochloric acid 2 mol/L in water
TCIAH1203-500ML
: TCI
:
- Pack type:Glass bottle
- Pk:500 mL
- Seq:2725
- Appearance:Colorless clear liquid
- Concentration (Neutralization Titration):1.9 to 2.1 mol/L
:
:14,04780
:8,III
:36.46 g/mol
:231-595-7
:1789
:GHS05
:GHS07
:Ambient
:7647-01-0
:HCl
:MFCD00011324