Order Entry
Belgium
ContactUsLinkComponent
Hydrochloric acid 1 mol/L in water
 
 
Hydrochloric acid 1 mol/L in water
  TCIAH1202-500ML
 :  TCI
 :  
undefined
 
 
Hydrochloric acid 1 mol/L in water
  TCIAH1202-500ML
 :  TCI
 :  

 

  • Pack type:
    Glass bottle
  • Pk:
    500 mL
  • Seq:
    2725

 

 

  • Appearance:
    Colorless clear liquid
  • Concentration (Neutralization titration):
    0.95 to 1.05 mol/L

 

 

 :
 :14,04780
 :8,III
 :36.46 g/mol
 :231-595-7
 :1789
 :GHS05
 :GHS07
 :Ambient
 :7647-01-0
 :HCl
 :MFCD00011324