2,5-Dimethoxytetrahydrofuran cis- and trans-mixture ≥98.0% (by GC)
Supplier: TCI
Certificates
About this item
CAS Number:696-59-3
:
Formula:C₆H₁₂O₃
MDL Number:MFCD00005359
:132.16 g/mol
Boiling Pt:146°C
Melting Pt:–45 °C
Density:1,021 g/cm³ (20 °C)
Flash Pt:35°C
Storage Temperature:Ambient
EINECS:211-797-1
UN:1993
ADR:3,III
:GHS02
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Colorless to Almost colorless clear liquid
- Purity (GC):min. 98.0 % (total of isomer)
Specification Test Results
Frequently Bought Together