


Calcium sulphate, Drierite™ 10-20 mesh, with indicator (cobalt (II) chloride)
:7778-18-9
:
:CaO₄S
:MFCD00010912
:136.14 g/mol
:1450 °C (dec.)
:Ambient
:231-900-3
:3077
:9,III
:GHS07
:GHS08
:GHS09
- Appearance:Blue Irregular granules
- Water:≤0.3 %
- Particle size:10 to 20 mesh
- Composition:≥98 % (CaSO4) Typical
Frequently Bought Together