Order Entry
Belgium
ContactUsLinkComponent
Calcium sulphate, Drierite™ 6 mm (4 mesh) for drying, with indicator (cobalt (II) chloride)
Irritant
Health
Environment
Calcium sulphate, Drierite™ 6 mm (4 mesh) for drying, with indicator (cobalt (II) chloride)
Certificates
undefined
Irritant
Health
Environment
Calcium sulphate, Drierite™ 6 mm (4 mesh) for drying, with indicator (cobalt (II) chloride)
Certificates

Specification Test Results

  • Appearance:
    Blue Irregular granules
  • Water:
    ≤0.3 %
  • Particle size:
    4 mesh
  • Composition:
    ≥98 % (CaSO4) Typical

Specification Test Results

Related Information

Frequently Bought Together