Sodium-1-decanesulphonate ≥98.0% (by titrimetric analysis) for ion pair chromatography
Supplier: TCI
Certificates
About this item
CAS Number:13419-61-9
:
Formula:C₁₀H₂₁NaO₃S
MDL Number:MFCD00007526
:244.33 g/mol
Melting Pt:> 300 °C
Storage Temperature:Ambient
EINECS:236-525-9
Specification Test Results
- Appearance:White powder to crystal
- Purity (Ion exchange titration):min. 98.0 %
- Absorbance (210nm):max. 1.000
- Absorbance (230nm):max. 0.300
- Absorbance (254nm):max. 0.130
Specification Test Results
Frequently Bought Together