Order Entry
Belgium
ContactUsLinkComponent
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99.0% (by HPLC, titration analysis) Good's buffer component for biological research
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99.0% (by HPLC, titration analysis) Good's buffer component for biological research
Supplier:  TCI
Certificates
undefined
Irritant
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS) ≥99.0% (by HPLC, titration analysis) Good's buffer component for biological research
Supplier:  TCI

Specification Test Results

  • Appearance:
    White to Almost white powder to crystal
  • Purity (HPLC):
    min. 99.0 area%
  • Purity (Nonaqueous Titration):
    min. 99.0 %
  • Absorbance:
    max. 0.15 (1mol/L、H2O、280 nm)
  • Solubility in Water:
    almost transparency

Specification Test Results

Related Information

Frequently Bought Together