Flat flange lids with 5 necks
Borosilicate glass 3.3.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
For use with SVL® reaction vessels. Available without, with two or five necks. Necks with GL thread.
Frequently Bought Together