Filtration crucibles
Supplier: SciLabware
Certificates
About this item
These filtration crucibles are made from Pyrex® borosilicate glass.
- With integral glass sintered disc
- Ideal for drying precipitates to constant mass at 110 °C
- Suitable for ignition work up to 450 °C