Universal bare fused-silica capillaries
Supplier: Agilent
Certificates
About this item
Universal Bare Fused-Silica Capillaries have a window, 750 mm effective length and 363 μm Ext.Ø, fitting into any CE instrument.
Frequently Bought Together
Universal Bare Fused-Silica Capillaries have a window, 750 mm effective length and 363 μm Ext.Ø, fitting into any CE instrument.