Order Entry
Belgium
Orders LinkContactUsLinkComponent
 

Multi-format Quatrorack systems

30 mm, Natural, 4/12/32, 50/15/1,5 ml
  GREI999251_1
Multi-format Quatrorack systems
  GREI999251_1
Multi-format Quatrorack systems

30 mm, Natural, 4/12/32, 50/15/1,5 ml

 
80,40 €
   
1 * 1 ST
 

Ideal for supporting 50 ml centrifuge tubes.

 
  • For tubes:
    50/15/1,5 ml
  • Hole Ø:
    30 mm
  • No. of Holes:
    4/12/32
  • Colour:
    Natural
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.