0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Lids
Microplate Lids
Lids for 96-well PCR plates, ThermoQuick
Lids for 96-well PCR plates, ThermoQuick
731-0676
:
Greiner Bio-One
Lids for 96-well PCR plates, ThermoQuick
731-0676
:
Greiner Bio-One
Colour:
Clear
Description:
Lid for Types 1 - 4 ThermoQuick microplates, transparent PC
Seq:
01
Pk:
50