Orders LinkContactUsLinkComponent
Inlet liners
Inlet liners
 :   Agilent
Inlet liners
 :  Agilent
Inlet liners
Inlet liners
These inlet liners prevent adsorption and ensure accurate sample transfer onto the GC column through a robust deactivated surface. These liners are recommended for trace level analysis of active analytes. Liners with inert wool retain non-volatiles, prolonging column life and lowering frequency of MS source maintenance.
  • Consistent high level of inertness
Product Family Options
Ultra inert liners
  • Description
    Ext.Ø
    Ø int.
  • Liner, GC, ultra inert, straight
    1 mm
    1 * 1 ST
    1 * 1 ST - 46,00 €
    46,00 €
    1 * 1 ST
       46,00 €
  • Liner, GC, ultra inert, straight, recommended for SPME (solid phase microextraction) injections
    0.75 mm
    1 * 1 ST
    1 * 1 ST - 59,00 €
    59,00 €
    1 * 1 ST
       59,00 €
  • Liner, GC, ultra inert, straight, splitless
    2 mm
    1 * 1 ST
    1 * 1 ST - 41,00 €
    41,00 €
    1 * 1 ST
       41,00 €
  • Liner, ultra inert deactivation, split, straight, glass wool
    4 mm
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • Liner, ultra inert deactivation, split, straight, glass wool
    4 mm
    1 * 25 ST
    1 * 25 ST - 793,00 €
    793,00 €
    1 * 25 ST
       793,00 €
  • Liner, ultra inert deactivation, splitless, single taper, glass wool
    4 mm
    1 * 5 ST
    1 * 5 ST - 224,00 €
    224,00 €
    1 * 5 ST
       224,00 €
  • Liner, ultra inert deactivation, splitless, single taper, glass wool
    4 mm
    1 * 25 ST
    1 * 25 ST - 971,00 €
    971,00 €
    1 * 25 ST
       971,00 €
  • Liner, ultra inert deactivation, splitless, single taper, no wool
    4 mm
    1 * 25 ST
    1 * 25 ST - 872,00 €
    872,00 €
    1 * 25 ST
       872,00 €
  • Liner, ultra inert deactivation, straight
    0.75 mm
    1 * 5 ST
    1 * 5 ST - 223,00 €
    223,00 €
    1 * 5 ST
       223,00 €
  • Liner, ultra inert deactivation, straight
    0.75 mm
    1 * 25 ST
    1 * 25 ST - 965,00 €
    965,00 €
    1 * 25 ST
       965,00 €
  • Liner, ultra inert, 2 mm, dimpled, splitless
    2 mm
    1 * 1 ST
    1 * 1 ST - 89,00 €
    89,00 €
    1 * 1 ST
       89,00 €
  • Liner, ultra inert, dimpled
    2 mm
    1 * 5 ST
    1 * 5 ST - 332,00 €
    332,00 €
    1 * 5 ST
       332,00 €
  • Liner, ultra inert, direct connect, dual taper, with hole
    4 mm
    1 * 5 ST
    1 * 5 ST - 425,00 €
    425,00 €
    1 * 5 ST
       425,00 €
  • Liner, ultra inert, direct connect, no hole
    4 mm
    1 * 5 ST
    1 * 5 ST - 405,00 €
    405,00 €
    1 * 5 ST
       405,00 €
  • Liner, ultra inert, direct connect, wool, bottom hole
    4 mm
    1 * 5 ST
    1 * 5 ST - 408,00 €
    408,00 €
    1 * 5 ST
       408,00 €
  • Liner, ultra inert, direct connect, wool, no hole
    4 mm
    1 * 5 ST
    1 * 5 ST - 411,00 €
    411,00 €
    1 * 5 ST
       411,00 €
  • Liner, ultra inert, direct connect, wool, top hole
    4 mm
    1 * 5 ST
    1 * 5 ST - 411,00 €
    411,00 €
    1 * 5 ST
       411,00 €
  • Liner, ultra inert, split, straight, glass wool
    4 mm
    1 * 100 ST
    1 * 100 ST - 2 200,00 €
    2 200,00 €
    1 * 100 ST
       2 200,00 €
  • Liner, ultra inert, split, straight, glass wool TouchLess package with non-stick O-ring
    4 mm
    1 * 1 ST
    1 * 1 ST - 49,00 €
    49,00 €
    1 * 1 ST
       49,00 €
  • Liner, ultra inert, splitless, double taper, no wool
    4 mm
    1 * 1 ST
    1 * 1 ST - 61,00 €
    61,00 €
    1 * 1 ST
       61,00 €
  • Liner, ultra inert, splitless, double taper, no wool
    4 mm
    1 * 5 ST
    1 * 5 ST - 223,00 €
    223,00 €
    1 * 5 ST
       223,00 €
  • Liner, ultra inert, splitless, single taper, glass wool
    4 mm
    1 * 100 ST
    1 * 100 ST - 2 690,00 €
    2 690,00 €
    1 * 100 ST
       2 690,00 €
  • Liner, ultra inert, splitless, single taper, glasswool TouchLess package with non-stick O-ring
    4 mm
    1 * 1 ST
    1 * 1 ST - 60,00 €
    60,00 €
    1 * 1 ST
       60,00 €
  • Liner, ultra inert, splitless, single taper, no wool
    4 mm
    1 * 100 ST
    1 * 100 ST - 2 420,00 €
    2 420,00 €
    1 * 100 ST
       2 420,00 €
  • Liner, ultra inert, splitless, single taper, no wool TouchLess package with non-stick O-ring
    4 mm
    1 * 1 ST
    1 * 1 ST - 54,00 €
    54,00 €
    1 * 1 ST
       54,00 €
  • Liner, ultra inert, splitless, singletaper, no wool
    4 mm
    1 * 5 ST
    1 * 5 ST - 201,00 €
    201,00 €
    1 * 5 ST
       201,00 €
  • Liner, ultra inert, universal, low pressure drop, glass wool
    4 mm
    1 * 5 ST
    1 * 5 ST - 279,00 €
    279,00 €
    1 * 5 ST
       279,00 €
  • Liner, ultra inert, universal, low pressure drop, glass wool
    4 mm
    1 * 25 ST
    1 * 25 ST - 1 210,00 €
    1 210,00 €
    1 * 25 ST
       1 210,00 €
  • Liner, ultra inert, universal, low pressure drop, glass wool
    4 mm
    1 * 100 ST
    1 * 100 ST - 3 350,00 €
    3 350,00 €
    1 * 100 ST
       3 350,00 €
  • Liner, ultra inert, universal, low pressure drop, GW TouchLess package with non-stick O-ring
    4 mm
    1 * 1 ST
    1 * 1 ST - 74,00 €
    74,00 €
    1 * 1 ST
       74,00 €
PTV liners
  • Description
    Ext.Ø
    Ø int.
  • Empty inserts/swirl holes
    1 * 10 ST
    1 * 10 ST - 304,00 €
    304,00 €
    1 * 10 ST
       304,00 €
  • Capillary inlet trap assembly
    1 * 1 ST
    1 * 1 ST - 365,00 €
    365,00 €
    1 * 1 ST
       365,00 €
  • PTV liner, single baffle, deactivated, 200 µl
    2 mm
    1 * 1 ST
    1 * 1 ST - 37,00 €
    37,00 €
    1 * 1 ST
       37,00 €
  • PTV liner, multi baffle, deactivated, 150 µl
    1,8 mm
    1 * 1 ST
    1 * 1 ST - 49,00 €
    49,00 €
    1 * 1 ST
       49,00 €
  • PTV liner, single baffle, glass wool, deactivated, 180 µl
    2 mm
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €
  • PTV liner, sintered glass, deactivated, 112 µl
    1,5 mm
    1 * 1 ST
    1 * 1 ST - 116,00 €
    116,00 €
    1 * 1 ST
       116,00 €
  • PTV liner, high temperature, quartz, 713 µl
    3,4 mm
    1 * 1 ST
    1 * 1 ST - 217,00 €
    217,00 €
    1 * 1 ST
       217,00 €
  • PTV liner, high temperature, borosilicate, 668 µl
    3,4 mm
    1 * 1 ST
    1 * 1 ST - 149,00 €
    149,00 €
    1 * 1 ST
       149,00 €
Split liners
  • Description
    Ext.Ø
    Ø int.
  • Split, straight liner with glass wool, non-deactivated, 990 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 36,00 €
    36,00 €
    1 * 1 ST
       36,00 €
  • Split, straight liner with glass wool, non-deactivated, 990 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 131,00 €
    131,00 €
    1 * 5 ST
       131,00 €
  • Split, straight liner with glass wool, non-deactivated, 990 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 569,00 €
    569,00 €
    1 * 25 ST
       569,00 €
  • Focus liner with glass wool, deactivated, 935 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 166,00 €
    166,00 €
    1 * 5 ST
       166,00 €
  • Focus liner, tapered with glass wool, deactivated, 880 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 218,00 €
    218,00 €
    1 * 5 ST
       218,00 €
  • Split, single taper liner with glass wool, deactivated, low pressure drop, 870 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 64,00 €
    64,00 €
    1 * 1 ST
       64,00 €
  • Split, single taper liner with glass wool, deactivated, low pressure drop, 870 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 239,00 €
    239,00 €
    1 * 5 ST
       239,00 €
  • Split, single taper liner with glass wool, deactivated, low pressure drop, 870 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 1 040,00 €
    1 040,00 €
    1 * 25 ST
       1 040,00 €
  • Split, single taper liner with glass wool, deactivated, 870 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 54,00 €
    54,00 €
    1 * 1 ST
       54,00 €
  • Split, single taper liner with glass wool, deactivated, 870 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 201,00 €
    201,00 €
    1 * 5 ST
       201,00 €
  • Split, single taper liner with glass wool, deactivated, 870 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 870,00 €
    870,00 €
    1 * 25 ST
       870,00 €
  • Split, straight liner with cup (for manual injections), 800 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 76,00 €
    76,00 €
    1 * 1 ST
       76,00 €
  • Split, straight liner with cup (for manual injections), 800 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 286,00 €
    286,00 €
    1 * 5 ST
       286,00 €
  • Split, straight liner with cup (for manual injections), 800 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 1 240,00 €
    1 240,00 €
    1 * 25 ST
       1 240,00 €
  • Split/splitless fast FocusLiner, with fused silica wool
    2,3 mm
    1 * 5 ST
    1 * 5 ST - 335,00 €
    335,00 €
    1 * 5 ST
       335,00 €
Splitless liners
  • Description
    Ext.Ø
    Ø int.
  • Splitless, single taper liner, deactivated, 900 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 49,00 €
    49,00 €
    1 * 1 ST
       49,00 €
  • Splitless, single taper liner, deactivated, 900 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 183,00 €
    183,00 €
    1 * 5 ST
       183,00 €
  • Splitless, single taper liner, deactivated, 900 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 791,00 €
    791,00 €
    1 * 25 ST
       791,00 €
  • Splitless, single taper liner, inert, 900 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 51,00 €
    51,00 €
    1 * 1 ST
       51,00 €
  • Splitless, single taper liner with glass wool, deactivated, 900 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 51,00 €
    51,00 €
    1 * 1 ST
       51,00 €
  • Splitless, single taper liner with glass wool, deactivated, 900 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 192,00 €
    192,00 €
    1 * 5 ST
       192,00 €
  • Splitless, single taper liner with glass wool, deactivated, 900 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 832,00 €
    832,00 €
    1 * 25 ST
       832,00 €
  • Splitless, straight liner, non-deactivated, 990 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 123,00 €
    123,00 €
    1 * 5 ST
       123,00 €
  • Splitless, straight liner, non-deactivated, 990 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 33,00 €
    33,00 €
    1 * 1 ST
       33,00 €
  • Splitless, double taper liner, deactivated, 800 μl
    4 mm
    1 * 1 ST
    1 * 1 ST - 56,00 €
    56,00 €
    1 * 1 ST
       56,00 €
  • Splitless, double taper liner, deactivated, 800 μl
    4 mm
    1 * 5 ST
    1 * 5 ST - 209,00 €
    209,00 €
    1 * 5 ST
       209,00 €
  • Splitless, double taper liner, deactivated, 800 μl
    4 mm
    1 * 25 ST
    1 * 25 ST - 905,00 €
    905,00 €
    1 * 25 ST
       905,00 €
  • Splitless, straight liner, deactivated, quartz, 250 µl
    2 mm
    1 * 1 ST
    1 * 1 ST - 44,00 €
    44,00 €
    1 * 1 ST
       44,00 €
  • Splitless, straight liner, deactivated, quartz, 250 μl
    2 mm
    1 * 25 ST
    1 * 25 ST - 720,00 €
    720,00 €
    1 * 25 ST
       720,00 €
  • Splitless, straight liner, deactivated, quartz, 250 μl
    2 mm
    1 * 5 ST
    1 * 5 ST - 166,00 €
    166,00 €
    1 * 5 ST
       166,00 €
  • Splitless, straight liner, non-deactivated, quartz, 250 μl
    2 mm
    1 * 25 ST
    1 * 25 ST - 754,00 €
    754,00 €
    1 * 25 ST
       754,00 €
  • Splitless, straight liner, non-deactivated, quartz, 250 μl
    2 mm
    1 * 5 ST
    1 * 5 ST - 174,00 €
    174,00 €
    1 * 5 ST
       174,00 €
  • Straight splitless liners, non deactivated, quartz, 250 μl
    2 mm
    1 * 1 ST
    1 * 1 ST - 46,00 €
    46,00 €
    1 * 1 ST
       46,00 €
Dimpled liners
  • Description
    Ext.Ø
    Ø int.
  • Dimpled, splitless, single taper liner, deactivated, 200 μl
    2 mm
    1 * 1 ST
    1 * 1 ST - 76,00 €
    76,00 €
    1 * 1 ST
       76,00 €
Direct connect liners
  • Description
    Ext.Ø
    Ø int.
  • Splitless, straight liner, non-deactivated, quartz, 140 μl
    1,5 mm
    1 * 1 ST
    1 * 1 ST - 35,00 €
    35,00 €
    1 * 1 ST
       35,00 €
  • Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl
    1,5 mm
    1 * 5 ST
    1 * 5 ST - 130,00 €
    130,00 €
    1 * 5 ST
       130,00 €
  • Straight, direct inlet liner, non-deactivated (for gas samples headspace or purge & trap), 140 μl
    1,5 mm
    1 * 25 ST
    1 * 25 ST - 562,00 €
    562,00 €
    1 * 25 ST
       562,00 €
  • Splitless, single taper, direct connect liner, Agilent proprietary deactivation
    4 mm
    1 * 1 ST
    1 * 1 ST - 103,00 €
    103,00 €
    1 * 1 ST
       103,00 €
  • Splitless, dual taper, direct connect liner, Agilent proprietary deactivation
    4 mm
    1 * 1 ST
    1 * 1 ST - 98,00 €
    98,00 €
    1 * 1 ST
       98,00 €
  • Splitless, single taper, direct connect liner, deactivated, inert
    4 mm
    1 * 1 ST
    1 * 1 ST
Liner O-rings
  • Description
    Ext.Ø
    Ø int.
  • Graphite O-ring
    9,63 mm
    6,35 mm
    1 * 10 ST
    1 * 10 ST - 94,00 €
    94,00 €
    1 * 10 ST
       94,00 €
  • Graphite O-ring, for splitless liner, temperatures above 350 °C
    9,63 mm
    6,52 mm
    1 * 10 ST
    1 * 10 ST - 94,00 €
    94,00 €
    1 * 10 ST
       94,00 €
  • O-ring for high temperature PTV
    1 * 1 ST
    1 * 1 ST - 133,00 €
    133,00 €
    1 * 1 ST
       133,00 €
  • Inlet liner non-stick O-ring
    1 * 10 ST
    1 * 10 ST - 23,00 €
    23,00 €
    1 * 10 ST
       23,00 €
  • Liner O-ring, non-stick flip-top
    1 * 10 ST
    1 * 10 ST - 36,00 €
    36,00 €
    1 * 10 ST
       36,00 €
Helix liners
  • Description
    Ext.Ø
    Ø int.
  • Helix liner, open ended, deactivated
    1 * 1 ST
    1 * 1 ST - 122,00 €
    122,00 €
    1 * 1 ST
       122,00 €
  • Helix liner, double taper, deactivated
    1 * 1 ST
    1 * 1 ST - 211,00 €
    211,00 €
    1 * 1 ST
       211,00 €
  • Helix liner, single taper, deactivated
    1 * 1 ST
    1 * 1 ST - 177,00 €
    177,00 €
    1 * 1 ST
       177,00 €
Aanbevelingen
Aanbevelingen worden alleen gepersonaliseerd op basis van uw winkelvoorkeuren als u daarvoor toestemming hebt gegeven door Verbeter mijn winkelervaring" op Mijn Account pagina" aan te zetten.
Anders ontvangt u algemene aanbevelingen.