Orders LinkContactUsLinkComponent
Filter plates, CHROMAFIL® MULTI 96
Supplier:   MACHEREY-NAGEL
Filter plates, CHROMAFIL® MULTI 96
Supplier:  MACHEREY-NAGEL
Certificates
Filter plates, CHROMAFIL® MULTI 96
Filter plates, CHROMAFIL® MULTI 96
11 Options Available Below
Product Details & Documents
These 96-well filter plates are used for simultaneous filtration of 96 samples. The plates are readily adaptable to all common automated robotic systems or to the CHROMABOND® MULTI 96 vacuum manifold (available as accessory).
  • Compatible 96-well polypropylene plates (monoblock)
  • Available with different membranes and pore sizes
  • Economical by saving time and solvent
Cellulose mixed esters: Hydrophilic membrane with very low adsorption for aqueous or polar solutions.
Regenerated cellulose (RC): Hydrophilic membrane for aqueous and organic-aqueous liquids, i.e. polar and medium polar sample solutions.
PTFE: Hydrophobic polytetrafluoroethylene membrane for nonpolar liquids and gases; very resistant towards all kinds of solvents as well as acids and bases.
PE: Polyethylene filter element with greater pore sizes for large particulate matter.
Glass microfibre (GF): Inert glass fibre filter with nominal pore size of 1 μm allows higher flow rates than small pore filters; for solutions with high loads of particulate matter or for highly viscous solutions (e.g., soil samples, fermentation broths).
Product Family Options
Product Information
  • Material
    Pore size
    Availability
    Price
  • MANA738770.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    Cellulose mixed esters
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738771.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    Cellulose mixed esters
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738656.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    Regenerated cellulose
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738657.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    Regenerated cellulose
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738660.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    PTFE
    0,20 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738661.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    PTFE
    0,45 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738662.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    PTFE
    1 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738663.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    PTFE
    3 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738659.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    PE
    40-100 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738655.2M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    Glass microfibre
    1 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
  • MANA738658.M
    CHROMAFIL® MULTI 96 filter plate
    CHROMAFIL® MULTI 96 filter plate
    Glass microfibre
    3 µm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,00 €
    38,00 €
    1 * 1 ST
       38,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • CHROMABOND® MULTI 96 vacuum manifold for 96-well monoblocks with reservoir tank, 3 spacer pairs, vacuum gauge and control valve
    CHROMABOND® MULTI 96 vacuum manifold for 96-well monoblocks with reservoir tank, 3 spacer pairs, vacuum gauge and control valve
    MANA738630.M
    Log in for availability
    1 * 1 KIT - 940,00 €