Orders LinkContactUsLinkComponent
Inlet septa
Inlet septa
Supplier:   Agilent
Inlet septa
Supplier:  Agilent
Certificates
Inlet septa
27 Options Available Below
Product Details & Documents
These septa are available for a variety of applications and have different upper temperature limits. The lower temperature septa are usually softer, seal better and can withstand more punctures (injections) than their high temperature counterparts.
  • Made up low bleed silicone rubber formulations
  • Efficient by using sharp syringe needles
  • Septa should be replaced to avoid leaks and sample loss
  • To be used with recommended temperature range
Product Family Options
Non stick bleed and temperature optimized (BTO) septa
Product Information
  • Ø
    Availability
    Price
  • HEWL5183-4757
    Septa, bleed and temperature optimised (BTO), non-stick
    Septa, bleed and temperature optimised (BTO), non-stick
    11 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 124,00 €
    124,00 €
    1 * 50 ST
       124,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4757
  • HEWL5183-4758
    Septa, bleed and temperature optimised (BTO), non-stick
    Septa, bleed and temperature optimised (BTO), non-stick
    5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 124,00 €
    124,00 €
    1 * 50 ST
       124,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4758
  • HEWL5183-4757-100
    Septa, bleed and temperature optimised (BTO), non-stick
    Septa, bleed and temperature optimised (BTO), non-stick
    11 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 228,00 €
    228,00 €
    1 * 100 ST
       228,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4757-100
Non stick advanced green septa
Product Information
  • Ø
    Availability
    Price
  • HEWL5183-4759
    Septa, Advanced Green, non-stick
    Septa, Advanced Green, non-stick
    11 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 114,00 €
    114,00 €
    1 * 50 ST
       114,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4759
  • HEWL5183-4760
    Septa, Advanced Green, non-stick
    Septa, Advanced Green, non-stick
    5 mm
    Log in for availability
    Log in for availability
    1 * 50 Pce
    1 * 50 Pce - 114,00 €
    114,00 €
    1 * 50 Pce
       114,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4760
  • HEWL5183-4759-100
    Septa, Advanced Green, non-stick
    Septa, Advanced Green, non-stick
    11 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 203,00 €
    203,00 €
    1 * 100 ST
       203,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4759-100
Non stick long life septa
Product Information
  • Ø
    Availability
    Price
  • HEWL5183-4761
    Septa, long-life, non-stick
    Septa, long-life, non-stick
    11 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 139,00 €
    139,00 €
    1 * 50 ST
       139,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4761
  • HEWL5183-4762
    Septa, long-life, non-stick
    Septa, long-life, non-stick
    5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 139,00 €
    139,00 €
    1 * 50 ST
       139,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4762
  • HEWL5183-4761-100
    Septa, long-life, non-stick
    Septa, long-life, non-stick
    11 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 249,00 €
    249,00 €
    1 * 100 ST
       249,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5183-4761-100
General purpose septa
Product Information
  • Ø
    Availability
    Price
  • HEWL5181-1260
    Septa, general purpose, grey
    Septa, general purpose, grey
    5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 48,00 €
    48,00 €
    1 * 25 ST
       48,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-1260
  • HEWL5181-1261
    Septa, general purpose, grey
    Septa, general purpose, grey
    5 mm
    Log in for availability
    Log in for availability
    1 * 25 ST
    1 * 25 ST - 48,00 €
    48,00 €
    1 * 25 ST
       48,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-1261
  • HEWL5181-1283-100
    Septa, general purpose, red
    Septa, general purpose, red
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-1283-100
  • HEWL5181-1283-50
    Septa, general purpose, red
    Septa, general purpose, red
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 87,00 €
    87,00 €
    1 * 50 ST
       87,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-1283-50
  • HEWL5080-8894-100
    Septa, general purpose, grey
    Septa, general purpose, grey
    11 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 156,00 €
    156,00 €
    1 * 100 ST
       156,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5080-8894-100
  • HEWL5080-8728-100
    Septa, general purpose, grey
    Septa, general purpose, grey
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 156,00 €
    156,00 €
    1 * 100 ST
       156,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5080-8728-100
  • HEWL5181-3383-50
    Septa, general purpose, red
    Septa, general purpose, red
    11 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 87,00 €
    87,00 €
    1 * 50 ST
       87,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-3383-50
  • HEWL5080-8896-50
    Septa, general purpose, grey
    Septa, general purpose, grey
    11 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 87,00 €
    87,00 €
    1 * 50 ST
       87,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5080-8896-50
  • HEWL5181-3383-100
    Septa, general purpose, red
    Septa, general purpose, red
    11 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 156,00 €
    156,00 €
    1 * 100 ST
       156,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-3383-100
  • HEWL5080-8728-50
    Septa, general purpose, grey
    Septa, general purpose, grey
    9,5 mm
    Log in for availability
    Log in for availability
    1 * 50 ST
    1 * 50 ST - 87,00 €
    87,00 €
    1 * 50 ST
       87,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5080-8728-50
Merlin microseal
Product Information
  • Ø
    Availability
    Price
  • HEWL5181-8815
    Microseal septum, stainless steel, rubber (30 psi)
    Microseal septum, stainless steel, rubber (30 psi)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 242,00 €
    242,00 €
    1 * 1 ST
       242,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-8815
  • HEWL5181-8816
    Merlin microseal kit, original low pressure system (includes nut and septum)
    Merlin microseal kit, original low pressure system (includes nut and septum)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 466,00 €
    466,00 €
    1 * 1 ST
       466,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-8816
  • HEWL5181-8833
    Merlin microseal kit, original low pressure system (includes nut and 2 septa)
    Merlin microseal kit, original low pressure system (includes nut and 2 septa)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 609,00 €
    609,00 €
    1 * 1 ST
       609,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-8833
  • HEWL5181-8839
    High sample volume septum kit (contains Merlin high pressure Microseal, six 23-gauge syringes, 500 vials and caps)
    High sample volume septum kit (contains Merlin high pressure Microseal, six 23-gauge syringes, 500 vials and caps)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 010,00 €
    1 010,00 €
    1 * 1 ST
       1 010,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5181-8839
  • HEWL5182-0853
    Microseal PTFE nut liners
    Microseal PTFE nut liners
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 13,00 €
    13,00 €
    1 * 2 ST
       13,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5182-0853
  • HEWL5182-3442
    High pressure Merlin microseal starter kit (includes microseal septum and nut)
    High pressure Merlin microseal starter kit (includes microseal septum and nut)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 489,00 €
    489,00 €
    1 * 1 ST
       489,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5182-3442
  • HEWL5182-3444
    Merlin microseal high pressure septum
    Merlin microseal high pressure septum
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 277,00 €
    277,00 €
    1 * 1 ST
       277,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5182-3444
  • HEWL5182-3445
    Microseal high pressure nut
    Microseal high pressure nut
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 253,00 €
    253,00 €
    1 * 1 ST
       253,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    HEWL5182-3445