Orders LinkContactUsLinkComponent
Basic Laboratory Chairs
Supplier:   bimos
Basic Laboratory Chairs
Supplier:  bimos
Certificates
Basic Laboratory Chairs
Basic Laboratory Chairs
8 Options Available Below
Product Details & Documents
The tried-and-tested all-rounder for use in laboratories is a great entry level laboratory chair, perfect for a range of workplace environments.
  • With a sturdy steel 5 star base
  • Available in two different frame colours: black or light grey
  • Choice of three different mechanisms: contact backrest, contact backrest with seat tilt adjustment and synchronous mechanism with weight regulation
Basic Laboratory can be supplied with Stamskin synthetic leather or with fabric upholstery. Not only is Stamskin Top easy to clean, but it also has antibacterial, antifungal and antimicrobial properties.
Conform to: DIN 68 877, German GS „safety tested“ certificate and Quality Management System DIN EN ISO 9001
 : Conform to: DIN 68 877, German GS 'safety tested' certificate and Quality Management System DIN EN ISO 9001
Product Family Options
Basic Laboratory 2, with castors
Product Information
  • Height
    Foot style
    Availability
    Price
  • BIMO9133-2571
    Basic laboratory, in stamskin top synthetic leather upholstery, black, with contact backrest, backrest height: 430 mm, black frame
    Basic laboratory, in stamskin top synthetic leather upholstery, black, with contact backrest, backrest height: 430 mm, black frame
    470 - 610 mm
    Castors
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 457,00 €
    457,00 €
    1 * 1 ST
       457,00 €
  • BIMO9134-2571
    Chair, with contact backrest
    Chair, with contact backrest
    470 - 610 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 522,00 €
    522,00 €
    1 * 1 ST
       522,00 €
  • BIMO9134-6902
    Basic laboratory 2 chair, in stamskin top synthetic leather upholstery, black, with contact backrest with tilting seat, backrest height: 530 mm, black frame
    Basic laboratory 2 chair, in stamskin top synthetic leather upholstery, black, with contact backrest with tilting seat, backrest height: 530 mm, black frame
    470 - 610 mm
    Castors
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 522,00 €
    522,00 €
    1 * 1 ST
       522,00 €
  • BIMO9134-6911
    Chair, with contact backrest
    Chair, with contact backrest
    470 - 610 mm
    Castors
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 522,00 €
    522,00 €
    1 * 1 ST
       522,00 €
  • BIMO9138-2571
    Basic laboratory 2 with synthetic leather upholstery, black, backrest height: 530 mm, black frame
    Basic laboratory 2 with synthetic leather upholstery, black, backrest height: 530 mm, black frame
    490 - 630 mm
    Castors
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 564,00 €
    564,00 €
    1 * 1 ST
       564,00 €
  • BIMO9138-6902
    Basic laboratory 2 with synthetic leather upholstery, blue, backrest height: 530 mm, black frame
    Basic laboratory 2 with synthetic leather upholstery, blue, backrest height: 530 mm, black frame
    490 - 630 mm
    Castors
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 564,00 €
    564,00 €
    1 * 1 ST
       564,00 €
Basic Laboratory 3, with glides and step
Product Information
  • Height
    Foot style
    Availability
    Price
  • BIMO9131-6801
    Basic laboratory 3 chair, in duotec fabric upholstery, black, with step and contact backrest, backrest height 430 mm, black frame
    Basic laboratory 3 chair, in duotec fabric upholstery, black, with step and contact backrest, backrest height 430 mm, black frame
    620 - 870 mm
    Glides
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 527,00 €
    527,00 €
    1 * 1 ST
       527,00 €
  • BIMO9131-6911
    Basic laboratory 3 chair, in skai® synthetic leather upholstery, grey, with step and contact backrest, backrest height: 430 mm, black frame
    Basic laboratory 3 chair, in skai® synthetic leather upholstery, grey, with step and contact backrest, backrest height: 430 mm, black frame
    620 - 870 mm
    Glides
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 527,00 €
    527,00 €
    1 * 1 ST
       527,00 €