Orders LinkContactUsLinkComponent
Phase separating paper
Phase separating paper
Supplier:   MACHEREY-NAGEL
Phase separating paper
Supplier:  MACHEREY-NAGEL
Certificates
Phase separating paper
22 Options Available Below
Product Details & Documents
These papers are made hydrophobic (impermeable to water) by impregnation with a silicone. With the aid of these filters, water can be separated from water-immiscible organic solvents in an elegant manner, by means of a simple filtration.
Specifications for Product Family
Specifications
  • Catalog No:
  • MANA430009
  • MANA430011
  • MANA430012
  • MANA430015
  • MANA430018
  • MANA430024
  • MANA484011
  • MANA484015
  • MANA484005
  • MANA484007
  • MANA484009
  • MANA484012
  • MANA484018
  • MANA484024
  • MANA484032
  • MANA584011
  • 516-6012
  • 516-6019
  • MANA584009
  • MANA584018
  • MANA584024
  • MANA584032
  • Weight:
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • 85 g/m²
  • Thickness:
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • 0,2 mm
  • Filtration speed:
  • 8 s (Herzberg)
  • 8 s (Herzberg)
  • 8 s (Herzberg)
  • 8 s (Herzberg)
  • 8 s (Herzberg)
  • 8 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
  • 27 s (Herzberg)
Specifications
Product Family Options
Grade MN 617 WA
Product Information
  • Grade
    Size
    Availability
    Price
  • MN 617 WA
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 20,50 €
    20,50 €
    1 * 100 ST
       20,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA430009
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    8 s (Herzberg)
  • MN 617 WA
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 26,60 €
    26,60 €
    1 * 100 ST
       26,60 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA430011
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    8 s (Herzberg)
  • MN 617 WA
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 32,30 €
    32,30 €
    1 * 100 ST
       32,30 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA430012
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    8 s (Herzberg)
  • MN 617 WA
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 41,90 €
    41,90 €
    1 * 100 ST
       41,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA430015
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    8 s (Herzberg)
  • MN 617 WA
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,10 €
    61,10 €
    1 * 100 ST
       61,10 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA430018
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    8 s (Herzberg)
  • MN 617 WA
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 98,40 €
    98,40 €
    1 * 100 ST
       98,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA430024
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    8 s (Herzberg)
Grade MN 616 WA
Product Information
  • Grade
    Size
    Availability
    Price
  • MN 616 WA
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 23,70 €
    23,70 €
    1 * 100 ST
       23,70 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484011
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 43,40 €
    43,40 €
    1 * 100 ST
       43,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484015
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    55 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 13,40 €
    13,40 €
    1 * 100 ST
       13,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484005
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    70 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 13,40 €
    13,40 €
    1 * 100 ST
       13,40 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484007
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 18,00 €
    18,00 €
    1 * 100 ST
       18,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484009
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 30,80 €
    30,80 €
    1 * 100 ST
       30,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484012
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 58,80 €
    58,80 €
    1 * 100 ST
       58,80 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484018
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 88,90 €
    88,90 €
    1 * 100 ST
       88,90 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484024
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    320 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 158,00 €
    158,00 €
    1 * 100 ST
       158,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA484032
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
Grade MN 616 WA, folded
Product Information
  • Grade
    Size
    Availability
    Price
  • MN 616 WA
    110 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,20 €
    61,20 €
    1 * 100 ST
       61,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA584011
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    150 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,20 €
    61,20 €
    1 * 100 ST
       61,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    516-6012
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    125 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,20 €
    61,20 €
    1 * 100 ST
       61,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    516-6019
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    90 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 61,20 €
    61,20 €
    1 * 100 ST
       61,20 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA584009
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    185 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 74,50 €
    74,50 €
    1 * 100 ST
       74,50 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA584018
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    240 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 113,00 €
    113,00 €
    1 * 100 ST
       113,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA584024
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)
  • MN 616 WA
    320 mm
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 182,00 €
    182,00 €
    1 * 100 ST
       182,00 €
    Specifications:
    More
    Specifications
    Cat. No.
    MANA584032
    Weight
    85 g/m²
    Thickness
    0,2 mm
    Filtration speed
    27 s (Herzberg)